Skip to Content
MilliporeSigma

918792

PIM-Norb-TB

Synonym(s):

Polymers of intrinsic microporosity

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C24H22N2)n
UNSPSC Code:
12162002
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Application

Polymers of intrinsic microporosity (PIMs) have generated significant interest for membrane separations and other applications since their initial discovery in 2004. PIMs are designed to have highly rigid and contorted backbone structures that cannot pack efficiently and thus exhibit microporosity. PIMs find applications in industry as they are solution processable, have a high permeability, and exhibit significant gas pair selectivities as membranes.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library