Skip to Content
MilliporeSigma

Skip To

GF03400322

Gilding metal alloy, Cu85Zn15

50m wire, 0.711mm diameter, as drawn

Synonym(s):

CU115200

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
$506.00

About This Item

NACRES:
NA.23
UNSPSC Code:
11101711

$506.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

wire

composition

Cu85Zn15

reaction suitability

core: copper, core: zinc

manufacturer/tradename

Goodfellow 034-003-22

L × diam.

50 m × 0.711 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF12027204GF52096215GF97415679
form

wire

form

wire

form

wire

form

wire

manufacturer/tradename

Goodfellow 034-003-22

manufacturer/tradename

Goodfellow 120-272-04

manufacturer/tradename

Goodfellow 520-962-15

manufacturer/tradename

Goodfellow 974-156-79

L × diam.

50 m × 0.711 mm

L × diam.

20 m × 0.711 mm

L × diam.

10 m × 0.711 mm

L × diam.

-

reaction suitability

core: copper, core: zinc

reaction suitability

core: copper, core: zinc

reaction suitability

core: copper, core: zinc

reaction suitability

core: copper, core: zinc


Still not finding the right product?

Explore all of our products under Gilding metal alloy, Cu85Zn15




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters