Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.23
UNSPSC Code:
11101712
form
annealed, foil (disks)
composition
Ni61Cr22Mo9Fe5
reaction suitability
core: chromium, core: iron, core: molybdenum, core: nickel
manufacturer/tradename
Goodfellow 034-997-35
size × thickness
4 mm × 0.2 mm
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation
Still not finding the right product?
Explore all of our products under Inconel® 625 - corrosion resistant alloy, Ni61Cr22Mo9Fe5
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.