Skip to Content
MilliporeSigma

GF04567192

Hastelloy® C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn

foil, 1m coil, 0.15mm thickness, annealed

Synonym(s):

NI140300

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
11101712
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

annealed, foil (coil)

composition

Ni57Mo17Cr16(Fe+W+Co)10

reaction suitability

core: chromium, core: cobalt, core: iron, core: molybdenum, core: nickel, core: tungsten

manufacturer/tradename

Goodfellow 045-671-92

size × thickness

1 m × 0.15 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Hastelloy is a registered trademark of Haynes International, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library