Skip to Content
MilliporeSigma

GF10040971

Platinum

wire reel, 0.1m, diameter 0.0006mm, wollaston, 99.9%

Synonym(s):

Platinum, PT005101

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Pt
CAS Number:
Molecular Weight:
195.08
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
11101803
MDL number:
eCl@ss:
26010603
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.9%

form

wire

manufacturer/tradename

Goodfellow 100-409-71

resistivity

10.6 μΩ-cm, 20°C

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow


Storage Class

13 - Non Combustible Solids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library