Skip to Content
MilliporeSigma

GF42150833

Polyethylene - Low Density

(LDPE), film, thickness 0.015 mm, L 5 m

Synonym(s):

ET311115

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100
Manufacturer/tradename:
Goodfellow 421-508-33
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

film

manufacturer/tradename

Goodfellow 421-508-33

L

5 m

thickness

0.015 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow


Still not finding the right product?

Explore all of our products under Polyethylene - Low Density


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library