Skip to Content
MilliporeSigma

GF43339664

Copper

foil, thickness 0.013 mm, 15 mm diameter, purity 99.99+%

Synonym(s):

CU000271

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Cu
CAS Number:
Molecular Weight:
63.55
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141711
MDL number:
eCl@ss:
38150101
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.99%, ±99.99%

form

foil

manufacturer/tradename

Goodfellow 433-396-64

resistivity

1.673 μΩ-cm, 20°C

diam.

15 mm

thickness

0.013 mm

diameter

15 mm

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

application(s)

metallization
perovskite solar cells

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow


Storage Class

13 - Non Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library