Skip to Content
MilliporeSigma

GF55152187

Polyvinylidenechloride

(PVDC), film, thickness 0.033 mm, size 150 × 150 mm

Synonym(s):

CV301230

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100
Manufacturer/tradename:
Goodfellow 551-521-87
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

film

manufacturer/tradename

Goodfellow 551-521-87

size

150 × 150 mm

thickness

0.033 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow


Still not finding the right product?

Explore all of our products under Polyvinylidenechloride


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library