Skip to Content
MilliporeSigma

GF64096263

Polyamide - Nylon 12

(PA 12), tube, outside diameter 0.63mm, L 2 m, wall thickness 0.065 mm

Synonym(s):

AM377006

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

tubes

manufacturer/tradename

Goodfellow 640-962-63

L

2 m

wall thickness

0.065 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow


Still not finding the right product?

Explore all of our products under Polyamide - Nylon 12


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library