Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.23
UNSPSC Code:
12352100
Manufacturer/tradename:
Goodfellow 650-820-01
form
film
manufacturer/tradename
Goodfellow 650-820-01
diam.
8 mm
thickness
0.09 mm
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
TPX is a registered trademark of Mitsui Petrochemical Industries, Ltd.
Still not finding the right product?
Explore all of our products under Polymethylpentene
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.