Skip to Content
MilliporeSigma

GF69005189

Cobalt-aluminum alloy, Co79Al21

powder, 150micron max. particle size, alloy pre-cursor, 500g

Synonym(s):

CO036010

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12352300
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

manufacturer/tradename

Goodfellow 690-051-89

weight

500 g

max. part. size

150 μm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library