Skip to Content
MilliporeSigma

GF72046302

Palladium

foil, thickness 0.60 mm, diameter 6 mm, purity 99.95%

Synonym(s):

PD000360

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141733
MDL number:
eCl@ss:
38191001
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 720-463-02

resistivity

9.96 μΩ-cm, 20°C

diam.

6 mm

thickness

0.60 mm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

application(s)

catalysts

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow


Still not finding the right product?

Explore all of our products under Palladium


Storage Class

13 - Non Combustible Solids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library