Skip to Content
MilliporeSigma

GF78986907

Invar® - Controlled Expansion Alloy, Fe64Ni36

sheet, 30 x 82mm, 9.5mm thickness, annealed

Synonym(s):

FE023230

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12352300
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

tubes

manufacturer/tradename

Goodfellow 789-869-07

size × thickness

30 x 82 mm × 9.5 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Invar is a registered trademark of Aperam Alloys Imphy


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library