Skip to Content
MilliporeSigma

GF93065362

Aluminum nitride

powder, 0.02 max. part. size (micron), weight 100 g, purity 99.5+%

Synonym(s):

AL646005

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12352311
eCl@ss:
38120507
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.5%

form

powder

manufacturer/tradename

Goodfellow 930-653-62

weight

100 g

max. part. size

0.02 (micron)

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow


Still not finding the right product?

Explore all of our products under Aluminum nitride




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library