Skip to Content
MilliporeSigma

GF96093153

Molybdenum

washer, size m6, length 25 pcs, outside diameter 11.57 - 12.00mm

Synonym(s):

MO00WA06

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
31161807
MDL number:
eCl@ss:
32150319
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 25

manufacturer/tradename

Goodfellow 960-931-53

resistivity

5.0 μΩ-cm, 20°C

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow


Storage Class

13 - Non Combustible Solids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library