Skip to Content
MilliporeSigma

21-1100

Ammonium nickel(II) sulfate hexahydrate

JIS special grade, ≥99.0%

Synonym(s):

Nickel ammonium sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(NH4)2Ni(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
394.99
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
239-793-5
MDL number:
Assay:
≥99.0%
Grade:
JIS special grade
Form:
crystalline
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

JIS special grade

assay

≥99.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: nickel

availability

available only in Japan

mp

85-89 °C (lit.)

SMILES string

N.N.[Ni++].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].OS(O)(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2H3N.Ni.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h2*1H3;;2*(H2,1,2,3,4);6*1H2/q;;+2;;;;;;;;/p-2

InChI key

ZZPAXHDZFDUTLY-UHFFFAOYSA-L



Still not finding the right product?


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library