Select a Size
Change View
About This Item
Linear Formula:
C10D7HO
CAS Number:
Molecular Weight:
151.21
NACRES:
NA.12
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
eCl@ss:
38190401
isotopic purity
97 atom % D
Quality Segment
form
solid
bp
278-280 °C (lit.)
mp
94-96 °C (lit.)
mass shift
M+7
SMILES string
[2H]c1c([2H])c([2H])c2c(O)c([2H])c([2H])c([2H])c2c1[2H]
InChI
1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H/i1D,2D,3D,4D,5D,6D,7D
InChI key
KJCVRFUGPWSIIH-GSNKEKJESA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact
Stable Isotopes Customer Service.
Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

