Skip to Content
MilliporeSigma

906441

QLAM-AβIδ1LVproR-U-13C Methyl Labeling Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.12
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

bio NMR: suitable

Quality Segment

shipped in

dry ice

storage temp.

−70°C

General description

QLAM-AβIδ1LVproR-U-[13C] Kit has 13C isotopomer precursors and contains protocol instructions for creation of isotopically-labeled proteins.

Application

For protein methyl group assignment by 13C isotope labeling of amino acid methyl groups separately or simultaneously.
Using QLAM-AβIδ1LVproR-U-[13C] kit, methyl bearing amino acids like alanine, isoleucine, leucine and valine are labeled with 13C isotopomer. This kit has been tested with protein isotopic labeling in E. coli. It is also used to provide good coverage across the structure of membrane proteins and information of structural dynamics and interactions in solid-state NMR spectroscopy.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact
Stable Isotopes Customer Service.



Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library