Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.43
UNSPSC Code:
41121808
Volume:
10 mL
Material:
borosilicate glass 3.3
Feature:
class AS
material
borosilicate glass 3.3
feature
class AS
packaging
pack of 1 ea
manufacturer/tradename
BRAND 23923
volume range
±0.02 mL
volume
10 mL
General description
The Modular Automatic Burette Concept! Fast to dismantle and easy to clean. Easy to repair - all individual components are replaceable! Class A/AS/AW glassware are calibrated to the highest-grade accuracy.
Application
Liquid handling, titration, volume measurements, dispensing
Features and Benefits
- Class AS
- DE-M marking Boro 3.3
- Detachable PTFE stopcock
- DIN EN ISO 385. Calibrated to deliver (TD, Ex)
- All BLAUBRAND® automatic burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Explore all of our products under BRAND® BLAUBRAND® compact-automatic burettes blue Schellbach stripe
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.