Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41121804
eCl@ss:
32039090
Material:
PMP
Packaging:
pack of 2 ea
material
PMP
feature
accuracy 0.5 mL Class B
packaging
pack of 2 ea
manufacturer/tradename
BRAND 36154
volume
500 mL
General description
With PP stopper. Error limits according to class B, DIN EN ISO 5215. Calibrated to contain (TC, In). Exposure to temperatures up to 60 ºC (autoclaving) will not cause permanently exceeded error limits! Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Explore all of our products under BRAND® Volumetric Flask
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.