Skip to Content
MilliporeSigma

CDS025378

(4-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}tetrahydro-2H-thiopyran-4-yl)acetic acid

AldrichCPR

Synonym(s):

4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]tetrahydro-2H-thiopyran-4-acetic acid

Select a Size

Change View

About This Item

Linear Formula:
C22H23O4N1S1
CAS Number:
UNSPSC Code:
12352106
NACRES:
NA.21
MDL number:
eCl@ss:
32150917
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


SMILES string

S1CCC(CC1)(NC(=O)OCC2c3c(cccc3)c4c2cccc4)CC(=O)O

InChI

1S/C22H23NO4S/c24-20(25)13-22(9-11-28-12-10-22)23-21(26)27-14-19-17-7-3-1-5-15(17)16-6-2-4-8-18(16)19/h1-8,19H,9-14H2,(H,23,26)(H,24,25)

InChI key

MOZSRIGQQHPMTM-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library