Skip to Content
MilliporeSigma

CDS025461

(3AS,8AS)-(+)-4,4,8,8-Tetrakis(3,5-di-i-propylphenyl)tetrahydro-2,2-dimethyl-6-phenyl-1,3-dioxolo[4,5-e]dioxaphosphepin

AldrichCPR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C61H81O4P1
CAS Number:
UNSPSC Code:
12352100
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


SMILES string

P2(OC([C@H]5OC(O[C@@H]5C(O2)(c7cc(cc(c7)C(C)C)C(C)C)c6cc(cc(c6)C(C)C)C(C)C)(C)C)(c4cc(cc(c4)C(C)C)C(C)C)c3cc(cc(c3)C(C)C)C(C)C)c1ccccc1

InChI

1S/C61H81O4P/c1-36(2)44-24-45(37(3)4)29-52(28-44)60(53-30-46(38(5)6)25-47(31-53)39(7)8)57-58(63-59(17,18)62-57)61(65-66(64-60)56-22-20-19-21-23-56,54-32-48(40(9)10)26-49(33-54)41(11)12)55-34-50(42(13)14)27-51(35-55)43(15)16/h19-43,57-58H,1-18H3/t57-,58-/m

InChI key

AYBKPJJNIOLFCE-YQOHNZFASA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library