Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.23
UNSPSC Code:
12352311
eCl@ss:
38120507
form
foil (disks - as rolled)
composition
Al97Mg3
reaction suitability
core: aluminum, core: magnesium
manufacturer/tradename
Goodfellow 207-190-54
size × thickness
8 mm × 0.2 mm
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Still not finding the right product?
Explore all of our products under Aluminum-magnesium alloy, Al97Mg3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.