Skip to Content
MilliporeSigma

GF26190884

Brass alloy, Cu63Zn37

foil, 25 x 25mm, 0.01mm thickness, hard, light tight (lt)

Synonym(s):

CU020208

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
30102010
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

light tight (lt)

form

foil (hard
hard)

composition

Cu63Zn37

reaction suitability

core: copper, core: zinc

manufacturer/tradename

Goodfellow 261-908-84

size × thickness

25 mm × 25 mm × 0.01 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow


Still not finding the right product?

Explore all of our products under Brass alloy, Cu63Zn37




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library