MilliporeSigma
Z679232
Microgard™ 2500 lab coat
size L
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.35
UNSPSC Code:
46181532
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
size
L
feature
CE compliant
color
white
General description
Microgard™ 2500 is designed to protect from saturating liquid spray and particulates, the fabric′s physical strength and flexibility ensure you are protected yet comfortable.
Features:
  • Collar
  • Stud front fastening
  • Left breast pocket
  • Lower right pocket
  • Bound seams
  • Designed as a single use/disposable product
Legal Information
Microgard is a trademark of Microgard Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)