Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C25H29NO10
CAS Number:
Molecular Weight:
503.50
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
eCl@ss:
25140904
grade
certified reference material
Quality Segment
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1 mg/mL (MeOH:H2O (50:50)(v/v))
format
single component solution
storage temp.
-10 to -25°C
General description
Naloxone-3beta-D-glucuronide is a major urinary metabolite of naloxone, an opiate drug used to counter the effects of opioid overdose. This certified reference solution is suitable as starting material for preparation of calibrators and controls in naloxone testing by LC/MS or GC/MS for forensic analysis, urine drug testing, or clinical toxicology applications.
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
WGK
WGK 2
Flash Point (°F)
75.2 °F
Flash Point (°C)
24 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


