Skip to Content
MilliporeSigma

P-162S

Phosphatidylethanol 16:0/18:2 (PEth 16:0/18:2) solution

1 mg/mL in methanol (as free phosphate), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Phosphatidylethanol 16:0/18:2 (PEth 16:0/18:2) solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C39H72O8P · NH4
CAS Number:
Molecular Weight:
718.00
UNSPSC Code:
12190000
eCl@ss:
32150319
Grade:
certified reference material
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol (as free phosphate)

format

single component solution

storage temp.

-65 to -96°C

SMILES string

CCOP(OC[C@H](OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)([O-])=O.[NH4+]

General description

Phosphatidylethanol 16:0/18:2 (PEth 16:0/18:2) is a direct biomarker of chronic and heavy alcohol consumption, formed only in the presence of ethanol. This Certified Spiking Solution® is suitable as starting material for preparing calibrators and controls for LC/MS-based methods in clinical toxicology, abstinence and treatment monitoring, forensic toxicology, and workplace or driving fitness testing. Ensure reliable calibration and QC standards with confidence and convenience using this Cerilliant® Second Source Lot certified reference material (CRM)—no special lot requests required.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library