Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C11H14N2O2
CAS Number:
Molecular Weight:
206.24
UNSPSC Code:
12352200
MDL number:
NACRES:
NA.24
grade
certified reference material
Quality Segment
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1 mg/mL in methanol
format
single component solution
storage temp.
-10 to -25°C
SMILES string
CCC(C(N)=O)(C(N)=O)C1=CC=CC=C1.O
InChI
1S/C11H14N2O2.H2O/c1-2-11(9(12)14,10(13)15)8-6-4-3-5-7-8;/h3-7H,2H2,1H3,(H2,12,14)(H2,13,15);1H2
InChI key
FZSBWHZDTUPYRX-UHFFFAOYSA-N
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
WGK
WGK 2
Flash Point (°F)
49.5 °F - closed cup
Flash Point (°C)
9.7 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


