Skip to Content
MilliporeSigma

T-019S

(-)-11-nor-9-Carboxy-Delta9-THC (Suitable for use with immunoassay) solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(−)-11-nor-9-Carboxy-Δ9-THC solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H28O4
CAS Number:
Molecular Weight:
344.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material

feature

second source lots, Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

application(s)

forensics and toxicology

format

single component solution

storage temp.

-10 to -25°C

SMILES string

CCCCCC(C=C1O)=CC2=C1[C@@]3([H])[C@@](C(C)(C)O2)([H])CCC(C(O)=O)=C3

InChI

1S/C21H28O4/c1-4-5-6-7-13-10-17(22)19-15-12-14(20(23)24)8-9-16(15)21(2,3)25-18(19)11-13/h10-12,15-16,22H,4-9H2,1-3H3,(H,23,24)/t15-,16-/m1/s1

InChI key

YOVRGSHRZRJTLZ-HZPDHXFCSA-N

General description

A second source analytical reference standard suitable in cannabinoid testing, clinical toxicology, forensic analysis, or urine drug testing by LC-MS/MS or GC/MS. Also known as THC acid, 11-nor-9-Carboxy-Δ9-THC is the main secondary metabolite of THC found in urine or blood of cannabis users.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library