Skip to Content
MilliporeSigma

T-050S

Temazepam glucuronide lithium salt solution

100 μg/mL in methanol (as free carboxylate), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Temazepam glucuronide lithium salt solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H20ClLiN2O8
Molecular Weight:
482.80
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material

feature

second source lots, Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free carboxylate)

application(s)

forensics and toxicology

format

single component solution

storage temp.

-10 to -25°C

SMILES string

[Li+].CN1C(=O)C(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C([O-])=O)N=C(c3ccccc3)c4cc(Cl)ccc14

InChI

1S/C22H21ClN2O8.Li/c1-25-13-8-7-11(23)9-12(13)14(10-5-3-2-4-6-10)24-19(20(25)29)33-22-17(28)15(26)16(27)18(32-22)21(30)31;/h2-9,15-19,22,26-28H,1H3,(H,30,31);/q;+1/p-1/t15-,16-,17+,18-,19?,22-;/m0./s1

InChI key

RQHXRNLYSZXQKI-NMJPLRNWSA-M

General description

The major urinary and blood metabolite of temazepam, an intermediate-acting 3-hydroxy benzodiazepine prescribed for the short-term treatment of sleeplessness in patients with difficulties remaining asleep. This new Certified Reference Standard is second source standard suitable for a variety of LCMS and GCMS applications, including forensic and forensic toxicology analysis, urine drug testing, and hydrolysis controls.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library