Skip to Content
MilliporeSigma

V-053

Thiamine pyrophosphate (Vitamin B1 pyrophosphate) solution

1 mg/mL (50:50 Methanol:Water), certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H19ClN4O7P2S
CAS Number:
Molecular Weight:
460.77
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (50:50 Methanol:Water)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[P](=O)([O-])(O[P](=O)(O)O)OCCc1[s]c[n+](c1C)Cc2c(nc(nc2)C)N.Cl

InChI

1S/C12H18N4O7P2S.ClH/c1-8-11(3-4-22-25(20,21)23-24(17,18)19)26-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H4-,13,14,15,17,18,19,20,21);1H

InChI key

YXVCLPJQTZXJLH-UHFFFAOYSA-N

General description

Thiamine pyrophosphate, also known as TPP or ThPP, is the biologically active form of vitamin B1. It is usually measured in whole blood or in plasma. While plasma thiamine concentration reflects recent intake rather than body stores, whole blood is the preferred specimen for thiamine assessment. This Certified Spiking Solution® is suitable for use as a starting material in preparation of linearity standards, calibrators, and controls in a variety of testing applications from diagnostic testing of thiamine deficiency to quality control testing of thiamine-based dietary supplements, fortified foods or pharmaceutical preparations.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

WGK

WGK 2

Flash Point (°F)

75.2 °F

Flash Point (°C)

24 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library