Skip to Content
MilliporeSigma

V-088

(6R,S)-5-Methyl-5,6,7,8-(6S)-5,6,7,8- Tetrahydrofolic acid calcium salt

vial of 5 mg, reference material, Cerilliant®

Synonym(s):

N-[4-[[(2-Amino-1,4,5,6,7,8-hexahydro-5-methyl-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H23CaN7O6
CAS Number:
Molecular Weight:
497.52
UNSPSC Code:
41104105
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

vial of 5 mg

manufacturer/tradename

Cerilliant®

format

neat

storage temp.

-65 to -96°C

InChI

1S/C20H25N7O6.Ca/c1-27-12(9-23-16-15(27)18(31)26-20(21)25-16)8-22-11-4-2-10(3-5-11)17(30)24-13(19(32)33)6-7-14(28)29;/h2-5,12-13,22H,6-9H2,1H3,(H,24,30)(H,28,29)(H,32,33)(H4,21,23,25,26,31);/q;+2/p-2/t12?,13-;/m0./s1

InChI key

VWBBRFHSPXRJQD-ZEDZUCNESA-L

General description

The bioavailability of natural folates is lower than for synthetic folic acid. USFDA′s new nutrition labeling 23 regulation of May, 2016 requires methods such as ISO TC34 that can estimate contents of folic acid and natural folates separately.
This method requires CRMs of Folic Acid, 5-CH3-THF, 5-CHO-THF, Folic acid triglutamate, Pte-Glu3 and the corresponding 13C labelled CRMs.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library