MilliporeSigma
1.17654
Transport Case
MAS-100® Sirius®
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 1 unit
manufacturer/tradename
Transport Case Sirius
technique(s)
: suitable using airmonitoring
application(s)
cosmetics
pharmaceutical
(Medical Device; Hospital)
compatibility
MAS-100® Sirius®
General description
The transport case is designed for one MAS-100 Sirius instrument. It has dedicated space for one Sirius air sampler equipped either with a perforated lid and a dust cover or a blind lid. There is as well space for a power supply and the USB cable.
The case shell is manufactured from PP, whereas the inlet is made from PE foam.
Legal Information
MAS-100 is a registered trademark of MBV AG
MAS-100 Sirius is a registered trademark of MBV AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)