MilliporeSigma
99100110
Opentrons® 50μL tip refill, filtered
Tips for AAW™ Workstation
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41121608
NACRES:
NB.02
Volume:
50 μL
Sterility:
sterile
Feature:
DNA free (Human), PCR clean, RNase and DNase free, autoclavable, endotoxin free, pyrogen free
Packaging:
pkg of 20 packs of 96 each
Manufacturer/tradename:
991-00110
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene
sterility
sterile
product line
AAW
feature
DNA free (Human), PCR clean, RNase and DNase free, autoclavable, endotoxin free, pyrogen free
manufacturer/tradename
991-00110
packaging
pkg of 20 packs of 96 each
volume
50 μL
General description
50 µL filtered pipette tips for the Opentrons Flex, provided sterile in racks or blister packs (refills). Tips are clear, non-conducting polypropylene tips, free of human DNA, PCR inhibitors, RNase/DNase, pyrogens, and endotoxins. Opentrons Flex Tips and Racks are autoclavable.
Legal Information
AAW is a trademark of Millipore Corporation
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Opentrons is a registered trademark of Opentrons Labworks, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)