MilliporeSigma
99100115
Opentrons Flex™ Heater-Shaker
AAW™ Accessory with adaptors and caddy
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41121521
NACRES:
NC.22
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
General description
The product is an accessory for use with the AAW™ Automated Assay Workstation. It allows for heating and shaking of plates on-deck. It comes with a caddy to adapt it to the deck, a calibration adapter and 4 plate adapters: Universal, Flat Bottom, PCR plate, and Deep Well plate. The heater is capable from ambient to 95°C, and programmable from 37°C to 95°C.
Other Notes
AAW™ Automated Assay Workstation (Cat. No. 99900256 or 99900241) is required for use of this product.
Legal Information
AAW is a trademark of Millipore Corporation
OPENTRONSFLEX is a trademark of Opentrons Labworks, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)