MilliporeSigma
99100300
Opentrons Flex™ Tough Auto-Sealing Lids
Opentrons Flex™ Tough Auto-Sealing Lids
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NC.02
Material:
polysulfone
Packaging:
pkg of 20 count
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polysulfone
product line
AAW
manufacturer/tradename
991-00300
packaging
pkg of 20 count
compatibility
Opentrons Flex Riser
General description
OPENTRONSFLEX™ Tough Auto-Sealing Lids provide automatic sealing of PCR plates on deck and are perfect for workflows that either want to minimize cross contamination or evaporation during their thermocycler incubations. The lids can be placed directly on the deck or with the OPENTRONSFLEX™ Riser, which will raise the position of the lids on the deck and make them stackable.The lids are added and removed from PCR plates using the AAW™ Flex Gripper and the OPENTRONSFLEX™ Thermocycler to create the seal. Once the incubation is complete, the customer can remove and dispose of the thermocycler lid and add a new one if there is a subsequent incubation.
Legal Information
OPENTRONSFLEX is a trademark of Opentrons Labworks, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)