Skip to Content
MilliporeSigma

MILLICL-TRIAL

Millicell® Cloud Subscription

Free Trial subscription to access Millicell® Cloud

Synonym(s):

Cloud application, Cloud storage

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32039090
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


General description

MILLICL-TRIAL offers free 6-month subscription to Millicell® Cloud features. MILLICL-1YR and MILLICL-5YR offer 1-year and 5-year subscription, respectively. Purchase Order of MILLICL-TRIAL is required to to activate the free trial subscription.

Application

Millicell® Cloud subscription adds Cloud connectivity to digital instruments, such as the Millicell® DCI, Millicell® ERS 3.0 Digital Voltohmmeter and Scepter™ 3.0 Handheld Cell Counter.

Features and Benefits

Data archiving, Plate maps for Millicell® ERS 3.0, Trend analysis, Cell Confluency analysis, Scepter™ histograms, Scepter™ experimental templates.

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany
Scepter is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Millicell® Cloud Subscription




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library