Skip to Content
MilliporeSigma

WTWA2AA112

pH 3110 Set 2, robust pH/mV meter

Synonym(s):

Hand held pH meter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41115603
eCl@ss:
32129202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library