Skip to Content
MilliporeSigma

10428392001

Roche

Pyrophosphatase, rec.

vial of 60 units, (Animal Origin Free, Antibiotic Free Production, Extended Impurity tested)

Synonym(s):

PPase, pyrophosphate phosphohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

(RNase-freein Glycerol)

packaging

vial of 60 units

manufacturer/tradename

Roche 10428392001

storage temp.

−20°C

General description

Recombinant Pyrophosphatase is an essential enzyme used in the in vitro transcription of mRNA. It catalyzes the hydrolysis of inorganic pyrophosphate (PPi), which is generated during RNA synthesis. By hydrolyzing PPi, the enzyme shifts the equilibrium of the mRNA synthesis reaction towards the formation of mRNA, thereby increasing the yield of the final product

Application

In Vitro Transcription: Recombinant Pyrophosphatase is primarily used in in vitro transcription systems to facilitate the synthesis of mRNA from DNA templates. Its role in hydrolyzing PPi is critical for maximizing RNA yield during transcription reactions.
Optimization of RNA Synthesis: The enzyme can be employed to optimize conditions for RNA synthesis, allowing researchers to achieve higher yields and better quality RNA for downstream applications.
Production of Modified RNA: By maintaining favorable reaction conditions, Recombinant Pyrophosphatase enables the incorporation of modified ribonucleoside triphosphates (NTPs) into RNA transcripts, which is important for studying the effects of modifications on RNA properties.
Functional RNA Studies: The enzyme is beneficial in producing high-quality RNA for functional studies, including RNA interference (RNAi), ribozymes, and RNA-based therapeutics, where yield and integrity are paramount.
RNA Labeling and Probing: Recombinant Pyrophosphatase can be utilized in protocols for labeling RNA or preparing RNA probes, enhancing the sensitivity and specificity of RNA detection techniques.

Features and Benefits

Scalable mRNA Synthesis: Designed for scalable mRNA production, suitable for both small-scale laboratory research and large-scale applications in biotechnology and pharmaceuticals.
Increased Yield: By hydrolyzing PPi, Recombinant Pyrophosphatase shifts the equilibrium towards mRNA synthesis, significantly increasing the yield of the final RNA product.
Enhanced Reaction Efficiency: The enzyme improves the efficiency of in vitro transcription reactions, allowing for more effective use of resources and time in RNA synthesis.
Versatile Applications: Recombinant Pyrophosphatase is applicable in various RNA synthesis and modification protocols, making it a valuable tool in molecular biology and biotechnology.
Improved RNA Quality: The enzyme’s action contributes to the production of high-quality RNA, essential for accurate and reliable downstream applications.


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

No data available

Flash Point (°C)

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library