Skip to Content

10436492001

Roche

CTP

vial of 25 μmol, (Animal Origin Free, Antibiotic Free Production, Extended Impurity tested)

Synonym(s):

Cytidine 5 -triphosphate, 5′-CTP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
41106305
NACRES:
NA.21
Form:
solution
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

packaging

vial of 25 μmol

manufacturer/tradename

Roche 10436492001

storage temp.

−20°C

General description

Cytidine triphosphate (CTP), along with adenosine triphosphate (ATP), )Guanosine-5′-triphosphate (GTP , uridine triphosphate (UTP), and potentially modified UTP, is part of the ribonucleotide mix required for synthesizing messenger RNA (mRNA) during transcription. This process occurs in the presence of RNA polymerase, which catalyzes the formation of RNA from the DNA template using these ribonucleotides.

Application

Guanosine-5′-triphosphate (GTP), cytidine triphosphate (CTP), adenosine triphosphate (ATP), and uridine triphosphate (UTP) are essential nucleotides involved in RNA synthesis. A significant application for these nucleotides is in vitro transcription (IVT) of mRNA, which is widely used in research and experimental studies.IVT kits incorporate these nucleotides to synthesize mRNA from DNA templates, enabling the production of RNA for various applications, including gene expression analysis and functional studies. However, off-the-shelf kits can restrict researchers′ ability to optimize the concentrations and ratios of these nucleotides according to the specific characteristics of different mRNA molecules, such as their composition and length. This limitation can impact the efficiency and quality of the synthesized mRNA. As a result, there is a growing interest in developing customizable approaches that allow for greater flexibility in reagent optimization, enhancing the performance of IVT reactions to better suit specific experimental requirements.

Features and Benefits

Scalable mRNA Synthesis: Designed for scalable mRNA production, suitable for both small-scale laboratory research and large-scale applications in biotechnology and pharmaceuticals.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library