Skip to Content
MilliporeSigma

0959-PA

(Glcα6)3β-Gly-PAA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.25
UNSPSC Code:
12352201
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


carrier

PAA carrier

product line

PA-Series

assay

95% (H-NMR)

form

lyophilized powder

mol wt

~20,000 Da

manufacturer/tradename

GlycoNZ

extent of labeling

20 mol% Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density

technique(s)

ELISA: suitable

probe type

trisaccharide

probe presentation

polymeric

storage temp.

−20°C

General description

PA-Series: Unlabeled Polyacrylamide Conjugates
This series of glycoconjugate probes consists of HOCH2CH2NH3+-salts for carboxylates and sulfates. PAA is poly[N-(2-hydroxyethyl) acrylamide], a flexible polymer chain which serves as an additional spacer. The spacer-arm for oligosaccharides is normally –(CH2)3–.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

Preparation Note

Purity: synthetic ω-aminoalkyl glycosides used for coupling with the polymer have a purity of >95% (HPLC and 1H-NMR)


Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library