Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C15H20NNaO5S
CAS Number:
Molecular Weight:
349.38
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
eCl@ss:
25140904
grade
analytical standard
Quality Segment
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
storage temp.
2-8°C
SMILES string
[Na+].Cc1cccc(C)c1N(CCSCC(O)C([O-])=O)C(=O)CO
InChI
1S/C15H21NO5S.Na/c1-10-4-3-5-11(2)14(10)16(13(19)8-17)6-7-22-9-12(18)15(20)21;/h3-5,12,17-18H,6-9H2,1-2H3,(H,20,21);/q;+1/p-1
InChI key
AVSNXORCABRXGJ-UHFFFAOYSA-M
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.