Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Grade:
HPLC grade
Technique(s):
HPLC: suitable
Bp:
81-82 °C
Vapor pressure:
72.8 mmHg
Signal Word
Danger
supp_hazards
Storage Class
3 - Flammable liquids
Flash Point (°F)
5.0 °F
Flash Point (°C)
-15 °C
PPE (personal protective equipment)
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system, Respiratory system
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



