product line
BioChemika
form
liquid
shelf life
limited shelf life, expiry date on the label
technique(s)
microbe id | staining: suitable
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
suitability
suitable for microscopy, spore forming microorganisms
SMILES string
[Cl-].[n+]2(c3c(nc4c2cc(c(c4)C)N)c(c(cc3)N)C)c1ccccc1
InChI
1S/C20H18N4.ClH/c1-12-10-17-19(11-16(12)22)24(14-6-4-3-5-7-14)18-9-8-15(21)13(2)20(18)23-17;/h3-11,22H,21H2,1-2H3;1H
InChI key
QRYAEWIQIBAZOJ-UHFFFAOYSA-N
General description
In Schaeffer-Fulton`s method, a primary stain-malachite green is forced into the spore by steaming the bacterial emulsion. Malachite green (Schaeffer & Fulton`s Spore Stain A: Cat. No. 90903) is water soluble and has a low affinity for cellular material, so vegetative cells may be decolorized with water. Vegetative cells are then counterstained with safarin. The components and the method are also available as kit (Cat. No. 04551).
Other Notes
Schaeffer & Fulton`s Spore Stain B: SafarinO 5 g/l in water
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
