Skip to Content
MilliporeSigma

42097

Mycobio Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Fungobiotic Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41171606
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shelf life

limited shelf life, expiry date on the label

composition

agar, 15.00 g/L , chloramphenicol, 0.05 g/L , cycloheximide, 0.50 g/L , dextrose, 10.00 g/L , papaic digest of soyabean meal, 10.00 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

6.5±0.2 (25 °C)

application(s)

microbiology

suitability

selective for Candida spp., selective for Trichophyton spp.

Application

Selective Agar for pathogenic fungi is recommended for the isolation of dermatophytes and many other pathogenic fungi.

Preparation Note

Suspend 35.55 grams in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. Distribute in tubes or flasks.
Sterilize by autoclaving at 15 lbs pressure (121°C) for 10 minutes. Cool the tubes in a slanted position. DO NOT REMELT OR OVERHEAT THE MEDIUM.
Warning: Cycloheximide is very toxic. Avoid skin contact or aerosol formation and inhalation.

Legal Information

NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Muta. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library