Skip to Content
MilliporeSigma

IA00007

DMG-PEG 2000

ultrapure grade

Synonym(s):

1,2-Dimyristoyl-rac-glycero-3-methoxypolyethylene glycol-2000, mPEG-2kDa-DMG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C122H242O50
CAS Number:
Molecular Weight:
2509.20
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

grade

ultrapure grade

assay

≥95% (,CAD, HPLC)

form

solid

packaging

pkg of 1 × 1 g (IA00007-1G)

application(s)

advanced drug delivery

lipid type

polymerizable lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCCCCCCCCCCCC)OCC(OC(CCCCCCCCCCCCC)=O)COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC

General description

DMG-PEG 2000, also known as dimyristoyl glycerophosphoethanolamine conjugated to polyethylene glycol 2000, is commonly used as a reagent to modify and functionalize biomolecules. It is often utilized to improve the solubility, stability, and pharmacokinetic properties of conjugated biomolecules, making it valuable in drug delivery, diagnostics, and biotechnology research.

Application

  • mRNA vaccine and therapeutic LNP formulation
  • siRNA and antisense oligonucleotide (ASO) delivery systems
  • Gene editing platform development (CRISPR/Cas9 and base editing)
  • Oncology and rare disease nucleic acid therapeutic research and development

Features and Benefits

  • Hydrophilic PEG2000 corona provides robust steric stabilization of LNP surfaceModulates
  • LNP particle size and polydispersity index (PDI) during formulation
  • Reduces opsonization and non-specific immune recognition in systemic circulation
  • Efficient intracellular delivery
  • DMG PEG2000 has broad compatibility with ionizable lipids, helper lipids, and cholesterol in standard LNP systems
  • It is extensively characterized and referenced across preclinical and clinical LNP literature
  • Research grade products that can easily be scaled to meets GMP needs.


Still not finding the right product?

Explore all of our products under DMG-PEG 2000


Flash Point (°C)

Not applicable

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library