Skip to Content
MilliporeSigma

NA-BE1-1

nBer e 1 (Bertholletia excelsa allergen 1)

pkg of 250 μg

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.21
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

assay

>95%

mol wt

Mw 12 kDa

packaging

pkg of 250 μg

manufacturer/tradename

InBio

format

neat

storage temp.

−20°C

General description

Source: Brazil Nuts
Formulation: Preservative and carrier-free in 1X PBS, pH 7.4. Filtered through 0.22μm filter.
Concentration: See product insert.
Purification: Purified from Pichia pastoris culture by multi-step chromatography. Purity >95 % by SDS-PAGE, under reducing conditions

Disclaimer

Allergens are provided for research and commercial use in vitro: not for human in vivo or therapeutic use.


Storage Class

12 - Non Combustible Liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library