Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
30 mg | Please contact Customer Service for Availability | $746.00 |
About This Item
Empirical Formula (Hill Notation):
C16H20N2O3
CAS Number:
Molecular Weight:
288.34
NACRES:
NA.24
$746.00
Please contact Customer Service for Availability
Quality Segment
grade
certified reference material, pharmaceutical secondary standard
agency
(Not traceable to any primary pharmacopeia standard)
API family
propranolol
CofA
current certificate can be downloaded
packaging
pkg of 30 mg
application(s)
pharmaceutical
storage temp.
-10 to -25°C
General description
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Analysis Note
These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
signalword
Warning
hcodes
Hazard Classifications
Carc. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
