Skip to Content
MilliporeSigma

PMC2006

XPhos

≥98%, ISOM8™

Sustainability features

Synonym(s):

XPhos, 2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl, 2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C33H49P
CAS Number:
Molecular Weight:
476.72
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352128
MDL number:
eCl@ss:
32151101
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

ISOM8™

Quality Segment

assay

(HPLC), ≥98%

form

solid

mol wt

476.72

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: C-C Bond Formation, reaction type: Cross Couplings, reaction type: Heck Reaction, reaction type: Hiyama Coupling, reaction type: Negishi Coupling, reaction type: Sonogashira Coupling, reaction type: Stille Coupling, reaction type: Suzuki-Miyaura Coupling, reagent type: ligand

greener alternative product score

old score: 2
new score: 1
Find out more about DOZN™ Scoring

sustainability

Sustainability features

mp

187-189  °C (Lit.), 187-190 °C (lit.)

application(s)

diagnostic assay manufacturing

functional group

phosphine

greener alternative category

storage temp.

room temp

SMILES string

CC(C)C1=CC(C(C)C)=CC(C(C)C)=C1C2=C(P(C3CCCCC3)C4CCCCC4)C=CC=C2

InChI

1S/C33H49P/c1-23(2)26-21-30(24(3)4)33(31(22-26)25(5)6)29-19-13-14-20-32(29)34(27-15-9-7-10-16-27)28-17-11-8-12-18-28/h13-14,19-25,27-28H,7-12,15-18H2,1-6H3

InChI key

UGOMMVLRQDMAQQ-UHFFFAOYSA-N

General description

Welcome to our ISOM8™ line of high-quality products available and able to scale for manufacturing. Within our ISOM8 team, we have expertise and documentation to support ISO 9001 industrial and non-GMP pharma production needs, as defined by our M-Clarity™ Program. Our portfolio of ISOM8 products and chemistries is ever evolving to support our valued Sigma-Aldrich® customers′ requirements. Please navigate to our documents section to look at quality documentation for this product, provided with support of our Elevate Program.

We are committed to bringing you Greener Alternative Products, which adhere to one of the  four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Atom Economy” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.

Application



XPhos, a powerful phosphine ligand developed by the developed by the Buchwald group, , significantly enhances palladium-catalyzed cross-coupling reactions for forming C-C, C-N, and C-O bonds, making it an indispensable tool in modern synthetic chemistry. Integration of XPhos into synthetic methodologies improves reaction efficiency, selectivity, and reliability while supporting both research-scale investigations and scalable manufacturing processes.
Notable Characteristics & Key Uses

  • Steric Effects: The bulky dialkyl biphenyl phosphine structure — featuring a 2,4,6-triisopropylphenyl flanking ring — provides steric protection that stabilizes the palladium center during catalytic processes and controls substrate approach and coordination.
  • Electronic Properties: XPhos exhibits strong sigma-donor properties, facilitating the formation of stable, highly reactive palladium complexes. Its well-balanced steric and electronic profile enhances catalytic activity and selectivity across diverse reaction conditions and substrate types.
  • Cross-Coupling Reactions: XPhos plays a pivotal role in Suzuki-Miyaura, Buchwald–Hartwig, Heck, and other palladium-catalyzed cross-coupling reactions, enabling efficient carbon–carbon and carbon–heteroatom bond formation.
  • Amination Reactions: Enables highly effective palladium-catalyzed amination of aryl halides and related substrates, providing direct access to aromatic amines and a broad range of nitrogen-containing compounds — including challenging primary and secondary amines.
  • Pharmaceutical & Agrochemical Synthesis: Due to its ability to facilitate the construction of complex molecular architectures, XPhos is widely employed in the synthesis of pharmaceutical compounds and agrochemicals, accelerating the development of new drug candidates.
  • Diverse Substrate Compatibility: XPhos accommodates a broad array of substrates, including sterically hindered electrophiles, aryl and heteroaryl halides, aryl chlorides, and functionalized nucleophiles, delivering consistently high yields where less capable ligands fall short.

Recent Studies and Innovations: 
  1. Stable active precatalyst formation: PdCl₂(XPhos)₂ serves as an easily accessible and efficient precatalyst for Suzuki–Miyaura cross-coupling of aryl/vinyl sulfonates and halides with broad functional group tolerance, including free OH, NH₂, and strongly coordinating pyridine. Its utility was further showcased in the synthesis of bioactive compounds dubamine (88%) and tamoxifen (78%). (Fatih Sirindil et al., 2022).
  2. XPhos in Controlled Pre-Catalyst Reduction for Cross-Coupling Reactions: XPhos was used an effective Buchwald-type ligand for the controlled in situ generation of active Pd(0) catalysts. Using PdCl₂(ACN)₂/XPhos in toluene/ethanol with K₂CO₃, complete and selective Pd(II)-to-Pd(0) reduction was achieved via alcohol oxidation (Mechanism A), preventing phosphine oxidation and boronate dimerization in Suzuki–Miyaura reactions. This practical, cost-effective, and green protocol is directly applicable to cross-coupling reactions in pharmaceutical and agrochemical synthesis. (Tommaso Fantoni et al., 2025).
Ligand for increased scope of Pd-catalyzed amination and amidation via arene sulfonates, aryl halides.

Legal Information

Usage subject to US Patents 6307087 and 6395916.
ISOM8 is a trademark of Sigma-Aldrich Co. LLC
M-CLARITY is a trademark of Merck KGaA, Darmstadt, Germany
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under XPhos


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library