Skip to Content
MilliporeSigma

PMC2015

XantPhos

≥98%, ISOM8™

Synonym(s):

Xantphos, 9,9-Dimethyl-4,5-bis(diphenyl­phosphino)xanthene, 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C39H32OP2
CAS Number:
Molecular Weight:
578.62
UNSPSC Code:
12352005
MDL number:
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

ISOM8™

Quality Segment

assay

≥98%

form

solid

purified by

HPLC

reaction suitability

reagent type: ligand, reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction, reagent type: ligand
reaction type: C-X Bond Formation, reagent type: ligand
reaction type: Cross Couplings, reagent type: ligand
reaction type: Hydroformylations, reagent type: ligand
reaction type: Miyaura Borylation Reaction, reagent type: ligand
reaction type: Stille Coupling, reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

224-228 °C

application(s)

diagnostic assay manufacturing
life science and biopharma

functional group

phosphine

storage temp.

room temp

SMILES string

CC1(c2c(Oc3c1cccc3P(c4ccccc4)c5ccccc5)c(P(c6ccccc6)c7ccccc7)ccc2)C

InChI

1S/C39H32OP2/c1-39(2)33-25-15-27-35(41(29-17-7-3-8-18-29)30-19-9-4-10-20-30)37(33)40-38-34(39)26-16-28-36(38)42(31-21-11-5-12-22-31)32-23-13-6-14-24-32/h3-28H,1-2H3

InChI key

CXNIUSPIQKWYAI-UHFFFAOYSA-N

General description

Welcome to our ISOM8™ line of high-quality products available and able to scale for manufacturing. Within our ISOM8 team, we have expertise and documentation to support ISO 9001 industrial and non-GMP pharma production needs, as defined by our M-Clarity™ Program. Our portfolio of ISOM8 products and chemistries is ever evolving to support our valued Sigma-Aldrich® customers′ requirements. Please navigate to our documents section to look at quality documentation for this product, provided with support of our Elevate Program.

Application

Xantphos is a widely utilized bidentate bisphosphine ligand celebrated for its exceptionally large natural bite angle (~111 degrees). This distinctive geometric property makes Xantphos highly effective for palladium-catalyzed cross-coupling, rhodium-catalyzed hydroformylation, and the formation of C-N and C-O bonds, establishing it as one of the most versatile ligands in modern organometallic catalysis.

Notable Characteristics & Key Uses

  • Chemical Structure: Xantphos features a rigid xanthene backbone providing a well-defined P-P distance. This structural rigidity allows it to coordinate metals in spanning or chelating modes, favoring both cis and trans complexes depending on reaction requirements.
  • Large Natural Bite Angle: The wide bite angle of Xantphos is its most distinctive feature, promoting reductive elimination and enabling selective catalytic pathways - particularly beneficial in hydroformylation and cross-coupling.
  • Bulky and Electron-Rich: The ligand′s steric profile and high electron density stabilize reactive metal complexes and enhance activity at the metal center.
  • Versatile Metal Coordination: Xantphos effectively coordinates palladium, rhodium, nickel, platinum, and ruthenium, offering broad utility across multiple catalytic systems.
  • Air and Moisture Stability: Xantphos is stable both as a solid and in solution, facilitating straightforward handling compared with more sensitive phosphine ligands.
  • Amination and Amidation: Highly effective for Buchwald-Hartwig amination and amidation, Suzuki-Miyaura coupling, and Negishi coupling to form C-N, C-O, and C-C bonds, with particular utility in heterocycle synthesis.
  • Carbonylation: Employed in palladium-catalyzed alkoxycarbonylation of alkenes for the efficient production of esters.
  • C-S and C-O Bond Formation: Applied in cross-couplings to construct C-S bonds (e.g., glycosyl thiol synthesis) and C-O bonds (e.g., benzylic arylation).
  • Hydrogenation: Utilized in ruthenium-catalyzed hydrogenation of carbon dioxide to formic acid or formate salts.
Recent Studies and Innovations 

  1. Synthesis of Anilines: Xantphos was deployed in a low-loading palladium-catalyzed process for the production of 3-bromo-2,5-difluoroaniline. The cross-coupling of 1,3-dibromo-2,5-difluorobenzene with benzophenone imine afforded the desired imine intermediate in high yield (Christoper D et al., 2025).
  2. Superior Performance in Alkyl-Alkyl Cross-Coupling: Xantphos has been demonstrated to outperform alternative bisphosphine ligands in alkyl-alkyl cross-coupling due to its larger bite angle and structural rigidity, both of which are critical determinants of catalytic efficiency (Magali et al., 2025).

Legal Information

ISOM8 is a trademark of Sigma-Aldrich Co. LLC
M-CLARITY is a trademark of Merck KGaA, Darmstadt, Germany
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under XantPhos


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library